siRNA Details
Gene Symbol: Gpam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008149.3 | NP_032175.2 | 13 | 1544 | ||
GenBank mRNA | AK047295.1 | - | 13 | 1545 | ||
AK137067.1 | - | 15 | 1825 | |||
AK173202.1 | - | 13 | 1569 | |||
BC019201.1 | - | 13 | 1549 | 5041120 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
