siRNA Details
Gene Symbol: Gpr12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010941.2 | NP_001010941.1 | 2 | 1406 | ||
NM_008151.3 | NP_032177.1 | 1 | 1192 | |||
GenBank mRNA | AK140298.1 | - | 2 | 1407 | ||
BC055746.1 | - | 1 | 1192 | 6415225 | ||
D21061.1 | - | 2 | 994 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
