siRNA Details
Gene Symbol: Cmklr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008153.3 | NP_032179.1 | 3 | 1323 | ||
GenBank mRNA | BC100612.1 | - | 3 | 1323 | 2654794 | |
U79525.1 | - | 3 | 1326 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
