siRNA Details
Gene Symbol: Gpr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008154.3 | NP_032180.1 | 2 | 570 | ||
GenBank mRNA | AK140302.1 | - | 2 | 541 | ||
AK140464.1 | - | 2 | 524 | |||
AK158917.1 | - | 2 | 533 | |||
BC138248.1 | - | 1 | 892 | 8861267 | ||
BC138250.1 | - | 1 | 892 | 8861269 | ||
D21062.1 | - | 1 | 945 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
