siRNA Details
Gene Symbol: Gpr33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008159.2 | NP_032185.1 | 2 | 614 | ||
GenBank mRNA | AF045766.1 | - | 2 | 637 | ||
BC064084.1 | - | 2 | 614 | 575313 | ||
GQ981323.1 | - | 2 | 655 | |||
GQ981327.1 | - | 2 | 717 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
