siRNA Details
Gene Symbol: Nmur1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010341.1 | NP_034471.1 | 2 | 902 | ||
GenBank mRNA | AF044602.1 | - | 2 | 902 | ||
BC120822.1 | - | 2 | 923 | 40129745 | ||
BC137775.1 | - | 2 | 923 | 8860794 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
