siRNA Details
Gene Symbol: Grcc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013535.1 | NP_038563.1 | 2 | 224 | ||
GenBank mRNA | AK009591.1 | - | 2 | 151 | ||
AK013899.1 | - | 2 | 210 | |||
BC005683.1 | - | 2 | 203 | 3154555 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
