siRNA Details
Gene Symbol: Grin2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008172.2 | NP_032198.2 | 9 | 2302 | ||
GenBank mRNA | AB154244.1 | - | 9 | 2188 | ||
AB154245.1 | - | 9 | 2188 | |||
D12822.1 | - | 9 | 2188 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
