siRNA Details
Gene Symbol: Gsc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010351.1 | NP_034481.1 | 3 | 1116 | ||
GenBank mRNA | X99239.1 | - | 3 | 1206 | ||
Y13149.1 | - | 3 | 1116 | |||
Y13150.1 | - | 3 | 1043 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
