siRNA Details
Gene Symbol: Gstm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010358.5 | NP_034488.1 | 8 | 803 | ||
GenBank mRNA | BC003822.1 | - | 8 | 626 | 3600444 | |
BC046758.1 | - | 8 | 656 | 5693552 | ||
BC091763.1 | - | 8 | 636 | 5102093 | ||
J03952.1 | - | 8 | 617 | |||
J04632.1 | - | 8 | 804 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
