siRNA Details
Gene Symbol: Gstt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010361.2 | NP_034491.2 | 3 | 914 | ||
GenBank mRNA | AK002392.1 | - | 3 | 444 | ||
AK158939.1 | - | 905 | ||||
BC012707.1 | - | 3 | 472 | 3994154 | ||
U48420.1 | - | 3 | 450 | |||
X98056.1 | - | 3 | 319 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
