siRNA Details
Gene Symbol: Gsto1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010362.2 | NP_034492.1 | 6 | 765 | ||
GenBank mRNA | AK027922.1 | - | 6 | 715 | ||
AK146834.1 | - | 6 | 727 | |||
AK168383.1 | - | 6 | 727 | |||
BC085165.1 | - | 6 | 703 | 30359900 | ||
U80819.1 | - | 6 | 765 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
