siRNA Details
Gene Symbol: Gzmk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008196.1 | NP_032222.1 | 5 | 736 | ||
GenBank mRNA | AB032200.1 | - | 981 | |||
BC117981.1 | - | 5 | 735 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
