siRNA Details
Gene Symbol: H1f0 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008197.3 | NP_032223.2 | 1 | 745 | ||
GenBank mRNA | AK051213.1 | - | 1 | 508 | ||
AK077473.1 | - | 1 | 422 | |||
AK090029.1 | - | 1 | 510 | |||
AK145675.1 | - | 1 | 397 | |||
AK146077.1 | - | 1 | 508 | |||
AK153990.1 | - | 1 | 509 | |||
AK155312.1 | - | 1 | 440 | |||
BC003830.1 | - | 1 | 494 | 3600538 | ||
BC011493.1 | - | 1 | 492 | 4166167 | ||
BC110361.1 | - | 1 | 497 | 5343233 | ||
X13171.1 | - | 1 | 475 |

More Information
Other Species Targeted

Gene Ontology Categories:
