siRNA Details
Gene Symbol: H2-Ab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207105.3 | NP_996988.2 | 2 | 233 | ||
GenBank mRNA | AK077436.1 | - | 2 | 221 | ||
AK170370.1 | - | 2 | 215 | |||
AY303784.1 | - | 1 | 66 | |||
AY303785.1 | - | 1 | 57 | |||
BY205244.1 | - | 2 | 252 | |||
M13539.1 | - | 2 | 148 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
