siRNA Details
Gene Symbol: H2-M9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008205.1 | NP_032231.1 | 1 | 52 | ||
GenBank mRNA | AF469488.1 | - | 1 | 52 | ||
AF539595.1 | - | 1 | 52 | |||
AK080256.1 | - | 1 | 76 | |||
BC106996.2 | - | 1 | 51 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
