siRNA Details
Gene Symbol: H2-DMb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010387.3 | NP_034517.2 | 2 | 427 | ||
GenBank mRNA | AK032745.1 | - | 2 | 428 | ||
BC003718.1 | - | 2 | 367 | 3486844 | ||
KJ650225.1 | - | 2 | 219 | |||
U20309.1 | - | 1 | 165 |
