siRNA Details
Gene Symbol: H2-DMb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010388.4 | NP_034518.1 | 2 | 238 | ||
GenBank mRNA | AK154923.1 | - | 2 | 342 | ||
BC052864.1 | - | 3 | 493 | |||
BC141105.1 | - | 2 | 304 | 9055717 | ||
BC145385.1 | - | 2 | 304 | 9053943 | ||
BC145386.1 | - | 2 | 304 | 9053944 | ||
BC171960.1 | - | 2 | 304 | 9054246 | ||
KJ650224.1 | - | 2 | 238 | |||
U20314.1 | - | 1 | 184 | |||
U20315.1 | - | 1 | 184 | |||
U20316.1 | - | 1 | 184 | |||
U20317.1 | - | 1 | 184 | |||
U35334.1 | - | 2 | 238 | |||
U35335.1 | - | 2 | 238 | |||
U35336.1 | - | 2 | 238 | |||
U35337.1 | - | 2 | 238 | |||
U35338.1 | - | 2 | 238 |
