siRNA Details
Gene Symbol: H2-Q1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010390.3 | NP_034520.2 | 8 | 1404 | ||
GenBank mRNA | BC028930.1 | - | 4 | 2239 | ||
BC080812.1 | - | 6 | 1333 | 6398887 | ||
U96752.1 | - | 8 | 1404 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
