siRNA Details
Gene Symbol: H2-Q2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010392.2 | NP_034522.2 | 5, 6 | 1012 | ||
GenBank mRNA | AY989880.1 | - | 5, 6 | 1012 | ||
BC138688.1 | - | 5, 6 | 1103 | 8861709 | ||
BC138689.1 | - | 5, 6 | 1103 | 8861711 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
