siRNA Details
Gene Symbol: H2-T23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010398.3 | NP_034528.1 | 2 | 162 | ||
GenBank mRNA | AB359230.2 | - | 2 | 162 | ||
AF057279.2 | - | 130 | ||||
AF082510.1 | - | 2 | 130 | |||
AF100699.1 | - | 2 | 103 | |||
AF100700.1 | - | 109 | ||||
AF100701.1 | - | 109 | ||||
AF100702.1 | - | 2 | 109 | |||
AK133502.1 | - | 2 | 162 | |||
BC099472.1 | - | 2 | 151 | 30842866 | ||
M11284.1 | - | 163 |
Gene Symbol: C920025E04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271005.1 | NP_001257934.1 | 2 | 149 | ||
GenBank mRNA | AK083376.1 | - | 2 | 162 | ||
AK083387.1 | - | 2 | 162 | |||
BG864910.1 | - | 2 | 148 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
