siRNA Details
Gene Symbol: Has3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008217.4 | NP_032243.2 | 4 | 1602 | ||
GenBank mRNA | AK028582.1 | - | 4 | 1546 | ||
BC138152.1 | - | 4 | 1498 | 8861171 | ||
BC138161.1 | - | 4 | 1497 | 8861180 | ||
U86408.2 | - | 4 | 1603 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
