siRNA Details
Gene Symbol: Hebp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013546.2 | NP_038574.2 | 3, 4 | 510 | ||
GenBank mRNA | AA022130.1 | - | 498 | |||
AB013095.1 | - | 3, 4 | 449 | |||
AF117613.1 | - | 3, 4 | 498 | |||
AK008919.1 | - | 3, 4 | 431 | |||
BC012654.1 | - | 3, 4 | 434 | 4216585 | ||
CT010262.1 | - | 3, 4 | 392 |
