siRNA Details
Gene Symbol: Hbegf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010415.2 | NP_034545.1 | 2 | 486 | ||
GenBank mRNA | AK052084.1 | - | 2 | 457 | ||
AK145548.1 | - | 2 | 455 | |||
AK160575.1 | - | 2 | 456 | |||
AK164309.1 | - | 2 | 487 | |||
BC089607.1 | - | 2 | 455 | 6751441 | ||
L07264.1 | - | 2 | 419 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
