siRNA Details
Gene Symbol: Hes1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008235.2 | NP_032261.1 | 2 | 412 | ||
GenBank mRNA | AK132536.1 | - | 2 | 423 | ||
AK133538.1 | - | 2 | 428 | |||
BC018375.1 | - | 2 | 412 | 4505427 | ||
BC051428.1 | - | 2 | 413 | 6478994 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
