siRNA Details
Gene Symbol: Hes3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008237.4 | NP_032263.2 | 3, 4 | 464 | ||
GenBank mRNA | AK090218.1 | - | 3, 4 | 465 | ||
BC116444.2 | - | 2, 3 | 379 | 40104666 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
