siRNA Details
Gene Symbol: Foxf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010426.2 | NP_034556.2 | 2 | 1091 | ||
GenBank mRNA | BC138805.1 | - | 2 | 1091 | 8861827 | |
BC138806.1 | - | 2 | 1091 | 8861828 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
