siRNA Details
Gene Symbol: Hint1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008248.2 | NP_032274.1 | 2 | 234 | ||
GenBank mRNA | AK002965.2 | - | 2 | 234 | ||
AK012433.1 | - | 2 | 240 | |||
BC070415.1 | - | 2 | 279 | 5714321 | ||
BC080296.1 | - | 2 | 218 | 6814185 | ||
U60001.1 | - | 2 | 204 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
