siRNA Details
Gene Symbol: Hipk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136065.2 | NP_001129537.1 | 15 | 3755 | ||
NM_001294143.1 | NP_001281072.1 | 15 | 3834 | |||
NM_001294144.1 | NP_001281073.1 | 15 | 3831 | |||
NM_010433.2 | NP_034563.2 | 15 | 3915 | |||
GenBank mRNA | AF077659.1 | - | 15 | 3678 | ||
AF170301.1 | - | 15 | 3584 | |||
AF170302.1 | - | 15 | 3665 | |||
AF208292.1 | - | 15 | 3668 | |||
AF273680.1 | - | 15 | 3555 | |||
AF333791.1 | - | 15 | 3458 | |||
AF333792.1 | - | 15 | 3542 | |||
BC031904.1 | - | 15 | 3567 | 5368577 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
