siRNA Details
Gene Symbol: Hnrnpk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301340.1 | NP_001288269.1 | 15 | 1381 | ||
NM_001301341.1 | NP_001288270.1 | 17 | 1508 | |||
NM_001301343.1 | NP_001288272.1 | 15 | 1367 | |||
NM_001301344.1 | NP_001288273.1 | 14 | 1309 | |||
NM_001301345.1 | NP_001288274.1 | 14 | 1309 | |||
NM_025279.3 | NP_079555.1 | 15 | 1381 | |||
GenBank mRNA | AK011428.1 | - | 15 | 1386 | ||
AK051313.1 | - | 14 | 1303 | |||
AK078777.1 | - | 17 | 1509 | |||
AK088462.1 | - | 14 | 1296 | |||
AK128891.1 | - | 15 | 1374 | |||
AK135519.1 | - | 15 | 1295 | |||
AK144245.1 | - | 8591 | ||||
AK150848.1 | - | 15 | 1361 | |||
AK151689.1 | - | 14 | 1263 | |||
AK152927.1 | - | 14 | 1263 | |||
AK160615.1 | - | 15 | 1351 | |||
AK160887.1 | - | 15 | 1383 | |||
AK166656.1 | - | 15 | 1386 | |||
AK166927.1 | - | 15 | 1339 | |||
AK167600.1 | - | 15 | 1372 | |||
AK168918.1 | - | 11 | 1284 | |||
AK169064.1 | - | 15 | 1371 | |||
BC006694.1 | - | 15 | 1348 | 3583037 | ||
BC089328.1 | - | 15 | 1304 | 6402614 | ||
EU604547.1 | - | 13 | 1162 |

More Information
Other Species Targeted

Gene Ontology Categories:
