siRNA Details
Gene Symbol: Hoxa11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010450.3 | NP_034580.1 | 2 | 845 | ||
GenBank mRNA | AK133481.1 | - | 2 | 845 | ||
BC052232.1 | - | 2 | 823 | |||
BC119302.1 | - | 2 | 780 | 8734051 | ||
U20370.1 | - | 2 | 801 |

More Information
Other Species Targeted

Gene Ontology Categories:
