siRNA Details
Gene Symbol: Hoxa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010451.2 | NP_034581.1 | 1 | 319 | ||
GenBank mRNA | AB221621.1 | - | 1 | 136 | ||
BC027688.1 | - | 1 | 271 | |||
BC117105.1 | - | 1 | 266 | 40126424 | ||
BC117107.1 | - | 1 | 266 | 40126426 | ||
M93292.1 | - | 1 | 230 | |||
M95599.1 | - | 1 | 319 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_012581, XM_008762936, M91802.
Gene Ontology Categories:
