siRNA Details
Gene Symbol: Hoxa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010452.3 | NP_034582.1 | 5 | 1607 | ||
GenBank mRNA | AK053218.1 | - | 1 | 749 | ||
AK076761.1 | - | 1 | 662 | |||
BC096612.1 | - | 4 | 1583 | 6476230 | ||
Y11717.1 | - | 5 | 1657 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
