siRNA Details
Gene Symbol: Hoxa6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010454.1 | NP_034584.1 | 1 | 143 | ||
GenBank mRNA | AB221546.1 | - | 1 | 143 | ||
BC119105.1 | - | 1 | 241 | 8733854 | ||
BC119107.1 | - | 1 | 241 | 8733856 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
