siRNA Details
Gene Symbol: Hoxa9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010456.3 | NP_034586.1 | 1 | 1716 | ||
GenBank mRNA | AB005457.1 | - | 1 | 1716 | ||
AB221551.1 | - | 1 | 450 | |||
BC055059.1 | - | 1 | 526 | 6529502 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
