siRNA Details
Gene Symbol: Hoxb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010461.2 | NP_034591.1 | 2 | 1569 | ||
GenBank mRNA | AB221615.1 | - | 2 | 508 | ||
AK131612.1 | - | 2 | 442 | |||
BC139209.1 | - | 2 | 749 | 8862231 | ||
BC139212.1 | - | 3 | 689 | 8862234 | ||
X13721.1 | - | 2 | 1568 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
