siRNA Details
Gene Symbol: Hoxc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008272.3 | NP_032298.1 | 3 | 1624 | ||
GenBank mRNA | AK012173.1 | - | 2 | 1012 | ||
AK078758.1 | - | 2 | 1050 | |||
BC050838.1 | - | 3 | 1621 | 6510837 | ||
X55318.1 | - | 2 | 1341 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
