siRNA Details
Gene Symbol: Hoxd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010467.2 | NP_034597.2 | 2 | 870 | ||
GenBank mRNA | AB221561.1 | - | 2 | 753 | ||
BC120537.1 | - | 2 | 769 | 40129460 | ||
BC120539.1 | - | 2 | 769 | 40129462 | ||
X60034.1 | - | 2 | 867 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001105884.
Gene Ontology Categories:
