siRNA Details
Gene Symbol: Hoxd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008273.2 | NP_032299.1 | 2 | 900 | ||
GenBank mRNA | AK054090.1 | - | 2 | 308 | ||
BC138115.1 | - | 2 | 900 | 8861134 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
