siRNA Details
Gene Symbol: Hoxd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010468.2 | NP_034598.2 | 2 | 663 | ||
GenBank mRNA | AK133705.1 | - | 2 | 531 | ||
AK138525.1 | - | 2 | 664 | |||
BC137716.1 | - | 1 | 288 | 8860735 | ||
BC137718.1 | - | 1 | 288 | 8860737 | ||
U56400.1 | - | 1 | 339 | |||
X73573.1 | - | 1 | 327 |
