siRNA Details
Gene Symbol: Lipc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008280.2 | NP_032306.2 | 4 | 724 | ||
GenBank mRNA | AK158285.1 | - | 4 | 575 | ||
AK158360.1 | - | 4 | 575 | |||
AY228765.1 | - | 4 | 476 | |||
BC021841.1 | - | 4 | 724 | 5124139 | ||
BC094050.1 | - | 4 | 511 | 5135403 | ||
X58426.1 | - | 4 | 495 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
