siRNA Details
Gene Symbol: Hspa8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031165.4 | NP_112442.2 | 4, 5 | 653 | ||
GenBank mRNA | AK004608.1 | - | 3, 4 | 942 | ||
AK035286.1 | - | 4, 5 | 651 | |||
AK075935.1 | - | 4, 5 | 650 | |||
AK081701.1 | - | 4, 5 | 652 | |||
AK098074.1 | - | 4, 5 | 650 | |||
AK145416.1 | - | 4, 5 | 573 | |||
AK145579.1 | - | 4, 5 | 647 | |||
AK146547.1 | - | 4, 5 | 654 | |||
AK146708.1 | - | 4, 5 | 648 | |||
AK146985.1 | - | 4, 5 | 651 | |||
AK147864.1 | - | 4, 5 | 651 | |||
AK149954.1 | - | 4, 5 | 648 | |||
AK150474.1 | - | 4, 5 | 645 | |||
AK150498.1 | - | 4, 5 | 645 | |||
AK150701.1 | - | 4, 5 | 655 | |||
AK150958.1 | - | 4, 5 | 648 | |||
AK151041.1 | - | 4, 5 | 651 | |||
AK151065.1 | - | 4, 5 | 651 | |||
AK151127.1 | - | 4, 5 | 654 | |||
AK151287.1 | - | 4, 5 | 651 | |||
AK151435.1 | - | 4, 5 | 651 | |||
AK151516.1 | - | 4, 5 | 657 | |||
AK151537.1 | - | 4, 5 | 648 | |||
AK151741.1 | - | 4, 5 | 651 | |||
AK151775.1 | - | 4, 5 | 653 | |||
AK151808.1 | - | 4, 5 | 654 | |||
AK151865.1 | - | 4, 5 | 651 | |||
AK151892.1 | - | 4, 5 | 653 | |||
AK151948.1 | - | 4, 5 | 654 | |||
AK151997.1 | - | 4, 5 | 651 | |||
AK152315.1 | - | 4, 5 | 654 | |||
AK152598.1 | - | 4, 5 | 651 | |||
AK152697.1 | - | 4, 5 | 651 | |||
AK152703.1 | - | 4, 5 | 651 | |||
AK152768.1 | - | 4, 5 | 654 | |||
AK152803.1 | - | 4, 5 | 654 | |||
AK153032.1 | - | 4, 5 | 653 | |||
AK153834.1 | - | 4, 5 | 651 | |||
AK157831.1 | - | 4, 5 | 651 | |||
AK159479.1 | - | 4, 5 | 651 | |||
AK162775.1 | - | 4, 5 | 651 | |||
AK163987.1 | - | 4, 5 | 651 | |||
AK164000.1 | - | 4, 5 | 651 | |||
AK166545.1 | - | 4, 5 | 653 | |||
AK166643.1 | - | 4, 5 | 665 | |||
AK166721.1 | - | 4, 5 | 648 | |||
AK166767.1 | - | 4, 5 | 651 | |||
AK166776.1 | - | 4, 5 | 654 | |||
AK166808.1 | - | 4, 5 | 651 | |||
AK166830.1 | - | 4, 5 | 649 | |||
AK166846.1 | - | 4, 5 | 651 | |||
AK166861.1 | - | 4, 5 | 654 | |||
AK166873.1 | - | 4, 5 | 651 | |||
AK166908.1 | - | 4, 5 | 654 | |||
AK166910.1 | - | 4, 5 | 651 | |||
AK166913.1 | - | 4, 5 | 651 | |||
AK166933.1 | - | 4, 5 | 651 | |||
AK167043.1 | - | 4, 5 | 653 | |||
AK167121.1 | - | 4, 5 | 654 | |||
AK167122.1 | - | 4, 5 | 651 | |||
AK167134.1 | - | 4, 5 | 649 | |||
AK167163.1 | - | 4, 5 | 653 | |||
AK167218.1 | - | 4, 5 | 651 | |||
AK167229.1 | - | 4, 5 | 652 | |||
AK167254.1 | - | 4, 5 | 652 | |||
AK167414.1 | - | 4, 5 | 651 | |||
AK167845.1 | - | 4, 5 | 654 | |||
AK167910.1 | - | 4, 5 | 654 | |||
AK168435.1 | - | 4, 5 | 654 | |||
AK168492.1 | - | 4, 5 | 653 | |||
AK168511.1 | - | 2, 3 | 189 | |||
AK168519.1 | - | 4, 5 | 651 | |||
AK168542.1 | - | 4, 5 | 653 | |||
AK168577.1 | - | 4, 5 | 651 | |||
AK168625.1 | - | 4, 5 | 651 | |||
AK168654.1 | - | 4, 5 | 651 | |||
AK168711.1 | - | 4, 5 | 651 | |||
AK168750.1 | - | 4, 5 | 654 | |||
AK168776.1 | - | 4, 5 | 654 | |||
AK168887.1 | - | 4, 5 | 653 | |||
AK168934.1 | - | 4, 5 | 654 | |||
AK169093.1 | - | 4, 5 | 651 | |||
AK169179.1 | - | 4, 5 | 651 | |||
AK169236.1 | - | 4, 5 | 651 | |||
AK169293.1 | - | 4, 5 | 665 | |||
AK169329.1 | - | 4, 5 | 654 | |||
AK169594.1 | - | 4, 5 | 650 | |||
AK171434.1 | - | 4, 5 | 652 | |||
AK178143.1 | - | 1, 2 | 132 | |||
AK184919.1 | - | 1, 2 | 132 | |||
AK190509.1 | - | 1, 2 | 132 | |||
AK192272.1 | - | 1, 2 | 132 | |||
AK193850.1 | - | 1, 2 | 132 | |||
AK193882.1 | - | 1, 2 | 132 | |||
AK195240.1 | - | 1, 2 | 132 | |||
AK198438.1 | - | 1, 2 | 132 | |||
AK199049.1 | - | 2, 3 | 359 | |||
AK199142.1 | - | 1, 2 | 132 | |||
AK199354.1 | - | 1, 2 | 132 | |||
AK199410.1 | - | 1, 2 | 165 | |||
AK199504.1 | - | 1, 2 | 132 | |||
AK200094.1 | - | 1, 2 | 132 | |||
AK200309.1 | - | 1, 2 | 132 | |||
AK200579.1 | - | 1, 2 | 132 | |||
AK201767.1 | - | 1, 2 | 132 | |||
AK202859.1 | - | 1, 2 | 132 | |||
AK203911.1 | - | 1, 2 | 132 | |||
AK204890.1 | - | 1, 2 | 132 | |||
AK205187.1 | - | 1, 2 | 132 | |||
AK206134.1 | - | 1, 2 | 132 | |||
AK206953.1 | - | 1, 2 | 132 | |||
AK209289.1 | - | 1, 2 | 132 | |||
AK210012.1 | - | 2, 3 | 198 | |||
AK211519.1 | - | 1, 2 | 132 | |||
AK212081.1 | - | 2, 3 | 196 | |||
AK212914.1 | - | 1, 2 | 152 | |||
AK214003.1 | - | 2, 3 | 179 | |||
AK216159.1 | - | 1, 2 | 132 | |||
BC006722.1 | - | 4, 5 | 675 | 3711399 | ||
BC066191.1 | - | 4, 5 | 607 | 30467052 | ||
BC085486.1 | - | 4, 5 | 639 | 30540914 | ||
BC089322.1 | - | 4, 5 | 632 | 30538640 | ||
BC089457.1 | - | 4, 5 | 610 | 30628505 | ||
BC094900.1 | - | 4, 5 | 567 | 30063483 | ||
BC106169.1 | - | 2, 3 | 331 | |||
BC106193.1 | - | 4, 5 | 635 | 6329283 | ||
M19141.1 | - | 4, 5 | 637 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
