siRNA Details
Gene Symbol: Hsd11b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044751.1 | NP_001038216.1 | 1 | 160 | ||
NM_008288.2 | NP_032314.2 | 2 | 186 | |||
GenBank mRNA | AI099306.1 | - | 1 | 163 | ||
AI182353.1 | - | 2 | 186 | |||
AK149518.1 | - | 1 | 158 | |||
AK165212.1 | - | 1 | 158 | |||
BC132364.1 | - | 2 | 126 | 40130641 | ||
BC138780.1 | - | 2 | 126 | 8861802 | ||
BC145415.1 | - | 2 | 126 | 9053973 | ||
DQ089001.1 | - | 1 | 62 | |||
S75207.1 | - | 1 | 107 | |||
X83202.1 | - | 2 | 189 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
