siRNA Details
Gene Symbol: Hsd3b5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008295.2 | NP_032321.2 | 4 | 1091 | ||
GenBank mRNA | BC012715.1 | - | 4 | 1095 | 4193717 | |
L41519.1 | - | 4 | 1076 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
