siRNA Details
Gene Symbol: Hspd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010477.4 | NP_034607.3 | 6 | 775 | ||
GenBank mRNA | AK088844.1 | - | 6 | 689 | ||
AK143882.1 | - | 6 | 680 | |||
AK146831.1 | - | 6 | 727 | |||
AK177219.1 | - | 3 | 200 | |||
AK179525.1 | - | 3 | 199 | |||
AK184625.1 | - | 3 | 206 | |||
AK187612.1 | - | 3 | 205 | |||
AK188312.1 | - | 3 | 199 | |||
AK216069.1 | - | 3 | 169 | |||
BC016400.1 | - | 6 | 661 | 3585270 | ||
BC106112.1 | - | 6 | 676 | |||
X53584.1 | - | 5 | 565 | |||
X55023.1 | - | 6 | 637 |

More Information
Other Species Targeted

Gene Ontology Categories:
