siRNA Details
Gene Symbol: Hspa1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010478.2 | NP_034608.2 | 1 | 2422 | ||
GenBank mRNA | AK159139.1 | - | 2390 | |||
AK165042.1 | - | 1 | 2429 | |||
M12573.1 | - | 1 | 943 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
