siRNA Details
Gene Symbol: Hspe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008303.4 | NP_032329.1 | 3, 4 | 522 | ||
GenBank mRNA | AK088121.1 | - | 3, 4 | 353 | ||
AK133876.1 | - | 3, 4 | 329 | |||
AK151935.1 | - | 3, 4 | 332 | |||
BC024385.1 | - | 3, 4 | 281 | 5004427 | ||
BC099385.1 | - | 3, 4 | 333 | 30848267 | ||
CJ238411.1 | - | 2, 3 | 206 | |||
U09659.1 | - | 3, 4 | 338 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
