siRNA Details
Gene Symbol: Htr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008313.4 | NP_032339.2 | 4 | 517 | ||
GenBank mRNA | AK141587.1 | - | 4 | 461 | ||
AK141625.1 | - | 4 | 518 | |||
JN418927.1 | - | 4 | 448 | |||
JN418928.1 | - | 4 | 434 | |||
JN418929.1 | - | 3 | 307 | |||
JN418930.1 | - | 3 | 323 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
