siRNA Details
Gene Symbol: Htr7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008315.2 | NP_032341.2 | 2 | 1043 | ||
GenBank mRNA | AK138150.1 | - | 2 | 845 | ||
AK139197.1 | - | 3 | 1103 | |||
AK139468.1 | - | 4 | 723 | |||
BC116986.1 | - | 2 | 1018 | 40126305 | ||
FM178515.1 | - | 2 | 936 | |||
FM178516.1 | - | 2 | 936 | |||
Z23107.1 | - | 2 | 1044 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
