siRNA Details
Gene Symbol: Slc6a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF013604.1 | - | 8, 9 | 1311 | ||
BC108978.1 | - | 9, 10 | 1480 | 40047534 | ||
BC108979.1 | - | 9, 10 | 1480 | 40047537 | ||
X66119.1 | - | 8, 9 | 1092 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
